Learn More
Applied Biosystems™ TaqMan™ Array Human Immune Panel
TaqMan™ Array Human Immune Panel
Brand:  Applied Biosystems™ 4370573
628.30 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.
Product Code. 11670099
  • £1030.00 / Each
Explore more special offers
Description
Description
The TaqMan™ Human Immune Array from Applied Biosystems™ is an efficient, easy-to-use micro fluidic card for quantitative gene expression analysis of targets known to have implications in immune response. This product is designed with 96 TaqMan™ Gene Expression Assays loaded in singletons into the array’s 384 wells and is compatible with an upgraded Applied Biosystems 7900HT Fast Real-Time PCR System. The targets include cytokines, chemokines, growth factors, immune regulators, apoptosis markers, ischemia markers, tissue-specific markers, and others including classic endogenous markers.This product is sold in 1 box of 4 cards.
The array is pre-formatted and inventoried, cost-effective, convenient and easy to use without needing expensive robotics. Results are reproducible and consistent across samples, studies, and labs —the same data quality from card-to-card and lot-to-lot — even with different operators.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Array Human Immune Panel
Human
Dried Down
MGB (Minor Groove Binder) (3'), Non-Fluorescent Quencher (3'), FAM (5')
TaqMan™
Standard
Cytokines, chemokines, growth factors, and other immune response genes
384-well Microfluidic Card
Each TaqMan™ Array well contains a single TaqMan™ Assay for one 2 μL reaction. The cards should be stored at 4°C.
TaqMan Assay for One 2 uL Reaction
FAM
4 cards
Cell Cycle & Proliferation, Apoptosis, Neurobiology, Signal Transduction, Biomarker-Related Pathway(s), Extracellular Matrix & Cell Adhesion, Reproduction, Toxicology & Drug Metabolism, Immunology, Oncology