missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ Array Human Inflammation

Product Code. 10658495 Shop All Applied Biosystems Products
Click to view available options
Quantity:
1 standard plate
Unit Size:
1 set
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10658495

Brand: Applied Biosystems™ 4414074

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

The panel of assays in this plate targets genes involved in a broad range of inflammatory diseases.

The genes fall into 4 classes: Channels (L-type calcium and Ligand gated); Enzymes and Inhibitors (Lipases, Kinases, Nitric Oxide Synthase, Phosphodiesterase, Prostaglandin Metabolism, and Proteases); Factors (Tumor necrosis factors, Nuclear factor Kappa-?, Interleukin, Annexins, Kinnogens); Receptors (GPCR, IL Receptor family, Adhesion molecules, TNF receptors and Nuclear receptors)

Gene Signature Plates are 96-well plates that are pre-configured with the most appropriate Applied Biosystems™ TaqMan Gene Expression Assays for a specific biological process, pathway, or disease state. Each plate contains predefined assays and endogenous controls dried-down in the wells, ready for accurate assessment of an entire gene signature in one simple experiment.

TaqMan Array Plates contain the same TaqMan Gene Expression Assays that have been available for years in a dried, 96-well format. Tests have found no difference in performance between TaqMan Assays supplied in the liquid format and assays dried into a TaqMan Array Plate. Ideal for projects such as validation of microarray and RNAi data, and pathway studies, the plates include enough TaqMan Assay in each well for one 20uL reaction. Each pack of TaqMan Array Plates is shipped with a CD containing the plate map along with the details of each TaqMan Assay. This information is imported into the real-time PCR instrument to enable proper labeling of data and easy access to Assay ID numbers for simple reordering.

  • Affordable; minimum order of three plates; ideal for small, medium or large-sized projects
  • Fast delivery; on your bench in days
  • Convenient; dried PCR primer and TaqMan™ probe sets supplied in a 96- well plate; just add master mix and your sample
  • TaqMan Array Plates have been optimized for use with TaqMan Gene Expression Master Mix, but are also compatible with TaqMan Universal PCR Master Mix
  • For studies using cultured cells, TaqMan Array Plates are also compatible (and recommended) with the TaqMan Gene Expression Cells-to-Ct™ Kit
  • TaqMan Array Plates must be run on a real-time PCR instrument with 96-well plate capability
  • he plates have been validated for use on Applied Biosystems™ 7000, 7300, 7500 and 7900HT Fast Real-Time PCR Systems using standard 96-well blocks, standard PCR cycling conditions, and 20μL reaction
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Each TaqMan™ Array Plate well contains a single TaqMan™ Assay for one 20 μL reaction. The plates can be stored at room temperature or at 4°C.
Form Dried Down
Format 96-well plate
Reaction Speed Standard
Internal Probe Modification MGB (Minor Groove Binder) (3'), Non-Fluorescent Quencher (3'), FAM (5')
Label or Dye FAM
Product Line TaqMan
Purification Method Standard
Quantity 1 standard plate
Research Category Signal Transduction, Oncology, Biomarker-Related Pathway(s), Extracellular Matrix & Cell Adhesion, Inflammation
Species Human
For Use With (Application) Gene Expression
No. of Reactions 1 x 20 μL Reactions
Show More Show Less
compliance-icons
Product Identifier
  • TaqMan Array Human Inflammation
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360-May damage fertility or the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Formamide

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.