missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ TaqMan™ Array, Human Insulin Receptor Pathway, Fast 96-well
TaqMan™ Array, Human Insulin Receptor Pathway, Fast 96-well
Brand: Applied Biosystems™ 4418798
98.94 GBP valid until 2024-11-30
Use promo code "21651" to get your promotional price.
This item is not returnable.
View return policy
Description
These 96-well plates are pre-configured with the most appropriate TaqMan™ Gene Expression Assays for a specific biological process, pathway, or disease state. Each plate contains predefined assays and endogenous controls dried-down in the wells, ready for accurate assessment of an entire gene signature in one simple experiment. The TaqMan™ Array Human Insulin Receptor Pathway 96-well Plate contains 92 assays to genes associated with Insulin Receptor pathways and 4 assays to candidate endogenous control genes.Specifications
Array Plate | |
Human | |
Dried Down | |
MGB (Minor Groove Binder) (3'), Non-Fluorescent Quencher (3'), FAM (5') | |
TaqMan™ | |
Fast |
96-array Plate | |
Each TaqMan™ Array Plate well contains a single TaqMan™ Assay for one 10 μL reaction. The plates can be stored at room temperature or at 4°C. | |
TaqMan Assay for One 10 uL Reaction | |
FAM | |
1 x 96-well plate | |
Cell Cycle & Proliferation, Endocrinology, Biomarker-Related Pathway(s), Extracellular Matrix & Cell Adhesion, Signal Transduction, Apoptosis, Oncology |
For Research Use Only. Not for use in diagnostic procedures.