missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ TaqMan™ Array Human Ligand Gated Ion Channels
96-well plate contains 92 assays to Ligand gated ion channel genes and 4 assays to candidate endogenous control genes
Brand: Applied Biosystems™ 4414092
Code : 50
Additional Details : Weight : 0.00450kg
Description
- The panel of assays in this plate targets genes in the 3 superfamilies of the ligand-gated channels:
- The Cys-loop receptors (GABAA receptor family, Glycine Receptor family, 5-HT Receptors/Serotonin, and Nicotinic acetylcholine receptor family)
- The Ionotropic glutamate receptors (AMPA, Kainate, NMDA)
- The ATP-gated Channels (P2X receptors)
- Also in the panel are genes which encode Ion channel family members
- Chloride channel
- TRP (transient receptor potential) cation channels
- Calcium Channel
- Sodium Channel
- Additionally one gene from the Cyclic nucleotide-gated channel and Potassium gated ion channel are also present
- Gene Signature Plates are 96-well plates that are pre-configured with the most appropriate TaqMan™ Gene Expression Assays for a specific biological process, pathway, or disease state
- Each plate contains predefined assays and endogenous controls dried-down in the wells, ready for accurate assessment of an entire gene signature in one simple experiment
- Affordable; minimum order of three plates; ideal for small, medium or large-sized projects
- Fast delivery; on your bench in days
- Convenient; dried PCR primer and TaqMan™ probe sets supplied in a 96- well plate; just add master mix and your sample
- Same TaqMan™Assay Quality, New Format
- TaqMan™ Array Plates contain the same high quality TaqMan™ Gene Expression Assays (PCR primers and TaqMan™ probe sets) that have been commercially available for years in a convenient, dried, 96-well format
- Extensive tests have found no difference in performance between TaqMan™Assays supplied in the traditional liquid format and assays dried into a TaqMan™ Array Plate.
Specifications
Array Plate | |
Human | |
Dried Down | |
MGB (Minor Groove Binder) (3'), Non-Fluorescent Quencher (3'), FAM (5') | |
TaqMan™ | |
Standard |
96-array Plate | |
Each TaqMan™ Array Plate well contains a single TaqMan™ Assay for one 20 μL reaction. The plates can be stored at room temperature or at 4°C. | |
TaqMan Assay for One 20 uL Reaction | |
FAM | |
Standard | |
Neurobiology |
For Research Use Only. Not for use in diagnostic procedures.