Learn More
Applied Biosystems™ TaqMan™ Array, Human NFKB, Fast 96-well
TaqMan™ Array, Human NFKB, Fast 96-well
Brand: Applied Biosystems™ 4418740
Code : 50
Additional Details : Weight : 0.00450kg
Description
These 96-well plates are pre-configured with the most appropriate TaqMan™ Gene Expression Assays for a specific biological process, pathway, or disease state. Each plate contains predefined assays and endogenous controls dried-down in the wells, ready for accurate assessment of an entire gene signature in one simple experiment. The panel of assays in the TaqMan™ Array 96-well Human NFkB Pathway Plate targets genes which encode the 5 proteins of the Rel⁄NFKB family: NFKB1, NFKB2, REL A, REL B, and REL. Also on the panel are genes from families that include: the IkB kinase; IkBs (inhibitor of kB); Toll-like receptor (TLR); Tumor Necrosis Factor (TNF); Tumor Necrosios Factor Receptors (TNFR); and Tumor Necrosis Factor Receptor Associated Factor (TRAF). Additional genes associated with NFKB function in apoptosis, immune⁄inflammation responses, as well as chemokines and cytokines are included.Specifications
Each TaqMan™ Array Plate well contains a single TaqMan™ Assay for one 10 μL reaction. The plates can be stored at room temperature or at 4°C. | |
96-well plate | |
MGB (Minor Groove Binder) (3'), Non-Fluorescent Quencher (3'), FAM (5') | |
TaqMan™ | |
Human | |
1 x 10 μL Reactions |
Dried Down | |
Fast | |
FAM | |
Immunology, Biomarker-Related Pathway(s), Extracellular Matrix & Cell Adhesion, Apoptosis | |
Gene Expression |
For Research Use Only. Not for use in diagnostic procedures.