Learn More
Applied Biosystems™ TaqMan™ Array, Human Nuclear Receptors, Fast 96-well
These 96-well plates are pre-configured with the most appropriate TaqMan™ Gene Expression Assays for a specific biological process, pathway, or disease state.
Brand: Applied Biosystems™ 4418753
Code : 50
Additional Details : Weight : 0.00450kg
Description
Each plate contains predefined assays and endogenous controls dried-down in the wells, ready for accurate assessment of an entire gene signature in one simple experiment. The TaqMan™ Array Human Nuclear Receptors 96-well Plate contains 92 assays to nuclear receptor associated genes and 4 assays to candidate endogenous control genes.
Specifications
Array Plate | |
Human | |
Dried Down | |
MGB (Minor Groove Binder) (3 in.), Non-Fluorescent Quencher (3 in.), FAM (5 in.) | |
TaqMan™ | |
Fast |
96-array Plate | |
Each TaqMan™ Array Plate well contains a single TaqMan™ Assay for one 10 μL reaction. The plates can be stored at room temperature or (at 4°C.) | |
TaqMan Assay for One 10 uL Reaction | |
FAM | |
1 x 96-well Plate | |
Cell Cycle & Proliferation, Apoptosis, Neurobiology, Signal Transduction, Oncology, Biomarker-Related Pathway(s), Extracellular Matrix & Cell Adhesion, Reproduction, Toxicology & Drug Metabolism, Immunology, Development |
For Research Use Only. Not for use in diagnostic procedures.