Learn More
Applied Biosystems™ TaqMan™ Array, Human Pathogenesis of ALS, Fast 96-well
TaqMan™ Array, Human Pathogenesis of ALS, Fast 96-well
Brand: Applied Biosystems™ 4418826
Code : 50
Additional Details : Weight : 0.00450kg
Description
These 96-well plates are pre-configured with the most appropriate TaqMan™ Gene Expression Assays for a specific biological process, pathway, or disease state. Each plate contains predefined assays and endogenous controls dried-down in the wells, ready for accurate assessment of an entire gene signature in one simple experiment. The TaqMan™ Array Human Pathogenesis of ALS 96-well Plate contains 28 assays to pathogenesis of ALS associated genes and 4 assays to candidate endogenous control genes. All assays are plated in triplicate.Specifications
Array Plate | |
Human | |
Dried Down | |
MGB (Minor Groove Binder) (3'), Non-Fluorescent Quencher (3'), FAM (5') | |
TaqMan™ | |
Cell Cycle & Proliferation, Apoptosis, Neurobiology, Biomarker-Related Pathway(s), Development, Immunology, Signal Transduction |
96-array Plate | |
Each TaqMan™ Array Plate well contains a single TaqMan™ Assay for one 10 μL reaction. The plates can be stored at room temperature or at 4°C. | |
TaqMan Assay for One 10 uL Reaction | |
FAM | |
Fast |
For Research Use Only. Not for use in diagnostic procedures.