Learn More
Applied Biosystems™ TaqMan™ Array, Human Transcription Factors (Non-Hox), Fast 96-well
TaqMan™ Array, Human Transcription Factors (Non-Hox), Fast 96-well
Brand:  Applied Biosystems™ 4418784
98.94 GBP valid until 2024-11-30
Use promo code "21651" to get your promotional price.
Product Code. 10752307
  • £194.00 / 1 set
Explore more special offers
For Research Use Only. All usage must comply with product instructions.
Description
Description
These 96-well plates are pre-configured with the most appropriate TaqMan™ Gene Expression Assays for a specific biological process, pathway, or disease state. Each plate contains predefined assays and endogenous controls dried-down in the wells, ready for accurate assessment of an entire gene signature in one simple experiment. The TaqMan™ Array Human Transcription Factors (Non-HOX) 96-well Plate contains 92 assays to Transcription Factors (Non-HOX) associated genes and 4 assays to candidate endogenous control genes.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Array Plate
Human
Dried Down
MGB (Minor Groove Binder) (3'), Non-Fluorescent Quencher (3'), FAM (5')
TaqMan™
Fast
96-array Plate
Each TaqMan™ Array Plate well contains a single TaqMan™ Assay for one 10 μL reaction. The plates can be stored at room temperature or at 4°C.
TaqMan Assay for One 10 uL Reaction
FAM
1 x 96-well plate
Cell Cycle & Proliferation, Apoptosis, Neurobiology, Signal Transduction, Gene Regulation, Development, Reproduction, Biomarker-Related Pathway(s), Immunology, Oncology