missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ TaqMan™ Array Mouse Immune Panel
Shop All Applied Biosystems Products
Click to view available options
Quantity:
4 cards
Unit Size:
Each
Description
The TaqMan™ Mouse Immune Array from Applied Biosystems™ is an efficient, easy-to-use micro fluidic card for quantitative gene expression analysis of targets known to have implications in immune response. This product is designed with 96 TaqMan™ Gene Expression Assays loaded in singletons into the array’s 384 wells and is compatible with an upgraded Applied Biosystems 7900HT Fast Real-Time PCR System. The targets include cytokines, chemokines, growth factors, immune regulators, apoptosis markers, ischemia markers, tissue-specific markers, and others including classic endogenous markers. This product is sold in 1 box of 4 cards.
The array is pre-formatted and inventoried, cost-effective, convenient and easy to use without needing expensive robotics. Results are reproducible and consistent across samples, studies, and labs —the same data quality from card-to-card and lot-to-lot — even with different operators.
The array is pre-formatted and inventoried, cost-effective, convenient and easy to use without needing expensive robotics. Results are reproducible and consistent across samples, studies, and labs —the same data quality from card-to-card and lot-to-lot — even with different operators.
Specifications
Specifications
| Type | Array Mouse Immune Panel |
| Format | 384-well Microfluidic Card |
| Species | Mouse |
| Content And Storage | Each TaqMan™ Array well contains a single TaqMan™ Assay for one 2 μL reaction. The cards should be stored at 4°C. |
| Form | Dried Down |
| Includes | TaqMan Assay for One 2 uL Reaction |
| Internal Probe Modification | MGB (Minor Groove Binder) (3'), Non-Fluorescent Quencher (3'), FAM (5') |
| Label or Dye | FAM |
| Product Line | TaqMan |
| Quantity | 4 cards |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction