Learn More
Applied Biosystems™ TaqMan™ Array Rat Endogenous Control Panel
TaqMan™ Array Rat Endogenous Control Panel
Brand:  Applied Biosystems™ 4378704
302.94 GBP valid until 2024-11-30
Use promo code "21651" to get your promotional price.
Product Code. 11620109
  • £594.00 / Each
Explore more special offers
Description
Description
The Applied Biosystems™ TaqMan™ Rat Endogenous Control Array is a 384-well micro flluidic card containing 16 rat TaqMan™ Gene Expression Assays commonly used as endogenous controls arrayed in triplicates. This product is compatible with an upgraded Applied Biosystems 7900HT Fast Real-Time PCR System. Endogenous Control Panels are sold in packs of 2.
The array is pre-formatted and inventoried, cost-effective, convenient and easy to use without needing expensive robotics. Results are reproducible and consistent across samples, studies, and labs —the same data quality from card-to-card and lot-to-lot — even with different operators.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TaqMan™ Array Rat Endogenous Control Panel
4°C
Control primer-probe set: TaqMan™ rat endogenous control array is sold in a pack of 2 cards and include a CD with assay information and an instrument software input file
384-well Microfluidic Card
Pharma and biopharma, real-time PCR (qPCR), real-time PCR-based gene expression profiling, target and lead identification and validation
2 Cards