missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ Array, Rat Inflammation, Fast 96-well

Product Code. 10408454 Shop All Applied Biosystems Products
Click to view available options
Quantity:
1 x 96-well plate
Unit Size:
1 set
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10408454

Brand: Applied Biosystems™ 4418726

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

TaqMan™ Array, Rat Inflammation, Fast 96-well

These 96-well plates are pre-configured with the most appropriate TaqMan™ Gene Expression Assays for a specific biological process, pathway, or disease state. Each plate contains predefined assays and endogenous controls dried-down in the wells, ready for accurate assessment of an entire gene signature in one simple experiment. The panel of assays in the TaqMan™ Array 96-well Rat Inflammation Plate targets genes involved in a broad range of inflammatory diseases. The genes fall into 4 classes: 1) Channels (L-type calcium and Ligand gated); 2) Enzymes and Inhibitors (Lipases, Kinases, Nitric Oxide Synthase, Phosphodiesterase, Prostaglandin Metabolism, and Proteases); 3) Factors (Tumor necrosis factors, Nuclear factor Kappa-Beta, Interleukin, Annexins, Kinnogens); and 4) Receptors (GPCR, IL Receptor family, Adhesion molecules, TNF receptors and Nuclear receptors).
TRUSTED_SUSTAINABILITY

Specifications

Type Array Plate
Format 96-array Plate
Species Rat
Content And Storage Each TaqMan™ Array Plate well contains a single TaqMan™ Assay for one 10 μL reaction. The plates can be stored at room temperature or at 4°C.
Form Dried Down
Includes TaqMan Assay for One 10 uL Reaction
Internal Probe Modification MGB (Minor Groove Binder) (3'), Non-Fluorescent Quencher (3'), FAM (5')
Label or Dye FAM
Product Line TaqMan
Quantity 1 x 96-well plate
Reaction Speed Fast
Research Category Inflammation, Signal Transduction
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.