missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ TaqMan™ Array, Standard, 96-well Plate; Format 8
TaqMan™ Array, Standard, 96-well Plate; Format 8
Brand: Applied Biosystems™ 4413266
106.75 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.
This item is not returnable.
View return policy
Description
Custom TaqMan™ Plates are cost-effective and convenient 96-well formats for TaqMan™ Gene Expression Assays. Choose from over 50,000 inventoried human, mouse, rat, canine, and rhesus macaque assays, and from five plate formats - single assays, or assays in duplicate or triplicate.Specifications
96-well Plate; Format 8 | |
Custom TaqMan™ Array Standard Plates, 20 μL rxns, 8-Well Plate Format, 12 Sample and 7 + 1 Controls in TaqMan™ Array Standard Plates for TaqMan™ Gene Expression Assays | |
Standard | |
Gene Expression, Genotyping |
Store at room temperature. | |
Standard | |
1 plate | |
20 μL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction