missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ Copy Number Assay
GREENER_CHOICE

Product Code. p-7076276 Shop All Applied Biosystems Products
Click to view available options
Quantity:
L (2900 reactions), made to order
M (750 reactions), made to order
S (360 reactions), made to order
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10478264

Brand: Applied Biosystems™ 4400291

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Applied Biosystems TaqMan Copy Number Assays use gold-standard TaqMan MGB probe chemistry to evaluate the copy number of genomic DNA targets using Applied Biosystems real-time PCR instruments and software.

Applied Biosystems TaqMan Copy Number Assays use gold-standard TaqMan MGB probe chemistry to evaluate the copy number of genomic DNA targets using Applied Biosystems real-time PCR instruments and software. TaqMan Copy Number Assays are run together with a TaqMan Copy Number Reference Assay in a duplex qPCR reaction; the copy number assay detects the target sequence, and the reference assay detects a sequence that is known to be present in two copies in the diploid genome. The results are then analyzed by the relative quantitation method using Applied Biosystems CopyCaller Software.

The predesigned TaqMan Copy Number Assay collection consists of human and mouse assay libraries for copy number variation (CNV) analysis. The human assay collection includes over 1.6 million assays targeting gene exons and introns, extragenic regions, and CNV sequences from the Database of Genomic Variants (DGV). The mouse collection includes over 180,000 assays targeting gene exons. Assays to common vector marker and reporter genes are also available for transgenic studies.

Benefits:

  • Specific—gold-standard TaqMan chemistry with minor groove binder (MGB) probes increase assay specificity by enabling shorter probes
  • Reproducible—robust assay designs developed with our bioinformatics pipeline enable accurate, reproducible, and reliable results
  • Easy to interpret—CopyCaller Software provides the calculated copy number and predicted copy number, along with confidence value and z-score quality metrics
  • Fast and simple—setup to primary analysis in 3–4 hours

Approximate ship time

4–6 days in North America and 6–10 days in Europe

All assay designs are the product of our bioinformatics pipeline, optimized over the course of more than a decade by leveraging manufacturing and assay performance data. TaqMan Assays have been cited in over 40,000 publications and are backed by more than 350 patents.

All of our predesigned TaqMan Assays are covered by the TaqMan Assays qPCR Guarantee.*

TaqMan Copy Number Reference Assays are sold separately.

Recommended master mix (sold separately): TaqMan Genotyping Master Mix (Cat. No. 4371355)

*Terms and conditions apply. For complete details, go to www.thermofisher.com/taqmanguarantee.

TRUSTED_SUSTAINABILITY

Specifications

3'Primer Modification None
5'Primer Modification None
Concentration 20X
Content And Storage 1 tube containing a 20X (S and M sizes) or 60X (L size) mix of pre-formulated assay (1 probe and 2 primers).

Store at -15 to -25°C.

Detection Method Primer-probe
Form Frozen
PCR Method qPCR
Reaction Speed Fast
For Use With (Equipment) 7500 System, 7500 Fast System, 7900HT System, StepOne™, StepOnePlus™, ViiA™ 7 System, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 7 Flex, QuantStudio 6 Pro, QuantStudio 7 Pro, QuantStudio™ 12k Flex, QuantStudio™ Absolute Q Digital PCR System
Green Features Sustainable packaging
Internal Probe Modification FAM (5'), MGB (Minor Groove Binder) (3')
Label or Dye FAM
Product Line TaqMan
Product Type Copy Number Assay
Purification Method Solid Phase Extraction (SPE)
Purity or Quality Grade RNase-Free, DNase-Free
Quantity S (360 reactions), made to order
Shipping Condition Room Temperature
Species Human
No. of Reactions 360 reactions
Volume 360 μL
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.