missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ TaqMan™ Copy Number Reference Assay, human, TERT
TaqMan™ Copy Number Reference Assay, human, TERT
£121.00 - £341.00
Specifications
3'Primer Modification | None |
---|---|
5'Primer Modification | None |
Content And Storage | Control Primer-Probe Set(s) |
Detection Method | Primer-probe |
Form | Frozen |
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
10035074
|
Applied Biosystems™
4403316 |
750 Reactions |
£121.00
Each |
Estimated Shipment: 29-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10157004
|
Applied Biosystems™
4403315 |
3000 Reactions |
£341.00
Each |
Estimated Shipment: 29-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Human TaqMan™ Copy Number Reference Assays are run with human TaqMan™ Copy Number Assays in a duplex real-time PCR reaction to detect and measure copy number variations (CNVs) and smaller regions in the human genome. Two options for genes that can be used as endogenous reference genes in humans are offered: RNase P and TERT. TaqMan™ Copy Number Reference Assays are designed to unique genomic sequences in the reference genome assembly (Build 37/hg19) and are required for relative quantitation of copy number targets.TaqMan™ Copy Number Assays quantitate the gene of interest and normalize to an endogenous reference gene known to be present in two copies in a diploid genome. Please note that TaqMan™ Copy Number Reference Assays are species specific.
TERT: The Alternative Reference Gene Option
TaqMan™ Copy Number Reference Assays have a VIC™ dye–labeled TAMRA™ probe and reference sequence–specific forward and reverse primers. The assays are not primer-limited.
TaqMan™ Copy Number Reference Assay RNase P is recommended as the standard reference assay for copy number analysis. This assay detects the Ribonuclease P RNA component H1 (H1RNA) gene (RPPH1) on chromosome 14, cytoband 14q11.2. The assay location is chr.14:20811565 on NCBI build 37. It has an 87 bp amplicon that maps within the single exon RPPH1 gene.
TaqMan™ Copy Number Reference Assay TERT is an alternative reference assay; it is recommended in the event that the RNase P assay functions poorly with a sample because of chromosomal aberrations or other issues. This assay targets the telomerase reverse transcriptase (TERT) gene located on chromosome 5, cytoband 5p15.33. The assay location is chr.5:1253373 on NCBI build 37. It has an 88 bp amplicon that maps within exon 16 of the TERT gene.
Simplest Copy Number Analysis Workflow
TaqMan™ Copy Number Assays have the simplest workflow of all currently available copy number analysis methods. The test assay (FAM™ dye–labeled), the reference assay (VIC™ dye–labeled), your sample DNA, and TaqMan™ Master Mix are combined and then run on an Applied Biosystems™ real-time PCR system using the standard TaqMan™ Copy Number Assay protocol. On average, setup to primary analysis takes only 3–4 hr (including an approximately 2 hour PCR run).
Powerful Data analysis Software
CopyCaller™ Software was developed specifically for TaqMan™ Copy Number Assay data analysis. This free, easy-to-use software utilizes a graphical interface that quickly calculates the possible copy numbers for a set of samples in a run. It also gives a confidence value for each copy number call, and has outlier functionality.
For Research Use Only. Not for human or animal therapeutic or diagnostic use.
Specifications
None | |
Control Primer-Probe Set(s) | |
Frozen | |
Low | |
Standard | |
Sustainable packaging | |
VIC | |
Configurable Assay | |
RNase-Free, DNase-Free | |
Human | |
20X |
None | |
Primer-probe | |
Tube | |
qPCR | |
TERT | |
TAMRA™ Quencher (3'), VIC™ (5') | |
TaqMan™ | |
HPLC | |
Dry Ice | |
Tert (Telomerase Reverse Transcriptase) |
For Research Use Only. Not for use in diagnostic procedures.