missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ Environmental Master Mix 2.0

Product Code. p-6833194 Shop All Applied Biosystems Products
Click to view available options
No. of Reactions:
200 Reactions
400 Reactions
800 Reactions
Unit Size:
1 set
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10575575

Brand: Applied Biosystems™ 4396838

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Accurate, real-time PCR-based pathogen detection in the presence of high levels of inhibitors.

Applied Biosystems™ TaqMan™ Environmental Master Mix 2.0 offers accurate, real-time PCR-based pathogen detection in the presence of high levels of inhibitors. Use this Master Mix to analyze environmental, food and other challenging samples.

  • Tailored for quantitative, real-time PCR experiments
  • Unrivalled sensitivity for challenging applications, low copy number target detection and multiplex PCR
  • Includes pathogen detection and viral⁄bacterial load quantification
  • Sensitive detection for reliable quantification of abundant and limited genetic targets
  • Stable mix for high-throughput handling
  • Upon receipt, store at -15°C to -25°C before first use, protected from light
  • After first use, store at 2°C to 8°C, protected from light

Optimized Formulation for Unrivalled Performance

TaqMan Environmental Master Mix 2.0 is a convenient 2X mix for target quantification that includes:

  • AmpliTaq Gold™ DNA Polymerase, UP (Ultra Pure), a highly purified DNA Polymerase for improved detection of bacterial targets. This hot-start enzyme is inactive at room temperature so reactions can be set up on the benchtop. Enzyme is activated during thermal cycling.
  • Passive internal reference based on proprietary ROX™ dye for precise data analysis.

For Research Use Only. Not for use in diagnostic procedures.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 2X
Content And Storage This TaqMan Environmental Master Mix 2.0 is a convenient 2X mix in a 5 mL bottle that includes: AmpliTaq Gold DNA Polymerase, UP (Ultra Pure), and a passive internal reference based on proprietary ROX dye for precise data analysis.
Upon receipt, store at -15 to -25°C before first use, protected from light. After first use, store at 2 to 8°C, protected from light.
Detection Method Primer-probe
Format Bottle
PCR Method qPCR
Polymerase AmpliTaq Gold DNA Polymerase
Reaction Speed Standard
For Use With (Equipment) 7000 System, 7300 System, 7500 Fast System, 7500 System, 7900HT Fast System, 7900HT System, StepOne™, Standard Mode
Passive Reference Dye ROX (Pre-mixed)
Product Line TaqMan
Product Type qPCR Master Mix
Purity or Quality Grade UP (Ultra Pure)
Quantity 200 reactions
Shipping Condition Dry Ice
Sufficient For 200 Reactions
For Use With (Application) Gene Expression, Genotyping, Pathogen Detection, Virus Detection
Hot Start Built-In Hot Start
No. of Reactions 200 Reactions
Sample Type Environmental Sample, Food
Volume 5 mL
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.