Learn More
Applied Biosystems™ TaqMan™ Exogenous Internal Positive Control Reagents
Shop All Applied Biosystems Products

Description
- Applied Biosystems™ TaqMan™ Exogenous Internal Positive Control Reagents contain a pre-optimized internal positive control (IPC) with pre-designed primers and TaqMan™ probe
- The IPC can be spiked into samples to distinguish true target negatives from PCR inhibition
- This kit allows users to amplify a low-copy target DNA in the same tube with the IPC
- Although the target and IPC DNAs may differ in initial copy number, the concentration of the IPC primers in the PCR reaction is limiting so that the amplification efficiency of the target reaction is not compromised
- Pre-designed primers and TaqMan™ probe eliminate assay design
- Rapid assay development guidelines can minimize optimization time Separate components provide flexibility in assay set-up
- Reagents may be stored at 2°C to 4°C if they are to be consumed within one month
- Otherwise, store at -20°C to -25°C
Note: This is not a quantitative kit and should not be used for normalization Instead, this kit is designed to distinguish types of negative results: a negative call for the target sequence and a positive call for the IPC indicate that no target sequence is present, and a negative call for the target sequence and negative call for the IPC suggest PCR inhibition.
Order Info
Note: This is not a quantitative kit and should not be used for normalization. Instead, this kit is designed to distinguish types of negative results: a negative call for the target sequence and a positive call for the IPC indicate that no target sequence
Specifications
Specifications
| Content And Storage | • 1 ml 10X Exo IPC Mix • 120 μl 10X Exo IPC Block • 200 μl 50X Exo IPC DNA Store at -20°C to -25°C. Reagents may be stored at 2–4°C if they are to be consumed within one month. |
| Format | Tube(s) |
| GC-Rich PCR Performance | Low |
| PCR Method | qPCR |
| Reaction Speed | Standard |
| Green Features | Sustainable packaging |
| Internal Probe Modification | TAMRA™ Quencher (3'), VIC™ (5') |
| Label or Dye | VIC |
| Product Line | TaqMan |
| Product Type | Exogenous Internal Positive Control Reagent |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.