missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ Gene Expression Assay, VIC primer-limited
GREENER_CHOICE

Product Code. 11907021 Shop All Applied Biosystems Products
Click to view available options
Quantity:
L (2900 reactions), made to order
M (750 reactions), made to order
S (360 reactions), made to order
Unit Size:
Each
This item is not returnable. View return policy
To order your assay via our online tool, please click here
 
For Research Use Only. All usage must comply with product instructions.
 

This Product Requires Additional Configuration

Please order Applied Biosystems™ TaqMan™ Gene Expression Assays through the Gene Expression Search Tool.

Product Code. 11907021

Brand: Applied Biosystems™ 4448485

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
To order your assay via our online tool, please click here
 
For Research Use Only. All usage must comply with product instructions.
 

This Product Requires Additional Configuration

Please order Applied Biosystems™ TaqMan™ Gene Expression Assays through the Gene Expression Search Tool.

Applied Biosystems TaqMan Gene Expression Assays, VIC primer-limited, are used for quantitative real-time PCR analysis of gene expression.

Applied Biosystems TaqMan Gene Expression Assays, VIC primer-limited, are used for quantitative real-time PCR analysis of gene expression and consist of a pair of unlabeled PCR primers and a TaqMan probe with a dye label (VIC) on the 5’ end and a minor groove binder (MGB) and nonfluorescent quencher (NFQ) on the 3’ end. These assays are primer-limited and ideal for multiplex reactions looking at two different gene targets in the same qPCR well.

Features include:
Easy to use—just add cDNA and master mix and run the qPCR—no melt curves required
Specific—TaqMan assays use proprietary MGB-containing probes that can be up to 15 bases shorter than non-MGB probes, improving the specificity of the assay
Sensitive—TaqMan assays are ideal for measuring low levels of expression or low-abundance targets
Accurate—identify small fold-changes with high accuracy of quantitation
Extensive content—over 1.8 million predesigned assays available for over 25 different species
Gold-standard TaqMan qPCR chemistry—TaqMan assays draw on Thermo Fisher Scientific's bioinformatics assay design pipeline to help ensure high specificity and minimal cross-reactivity, even for gene variants with high sequence homology
Ideal for multiplexing—combine one FAM-labeled and one VIC-labeled assay to create a duplex assay for two different gene targets in the same qPCR well
Ideal for control assays—use a VIC primer-limited assay for high-expressing endogenous control genes

Approximate ship time
Made to order: 4–6 days in North America, 6–10 days in Europe

Pre-formulated assay
• 2 unlabeled PCR primers (150 nM each final 1X concentration)
• 1 VIC dye-labeled TaqMan MGB probe (250 nM final 1X concentration)

TaqMan Gene Expression assays are the gold standard in real-time PCR gene expression studies, built on more than 20 years of experience. Each assay includes target primers and a sequence-specific probe optimized for the best functional performance. No additional design, optimization, or melt curve analysis is needed. Available in a wide variety of formats and species, new assay designs are constantly added to help meet your research needs. TaqMan assays have been cited in over 40,000 publications and are backed by more than 350 patents. All of our predesigned TaqMan Gene Expression assays are covered by the TaqMan Assays qPCR Guarantee.*

Recommended master mix: TaqMan Fast Advanced Master Mix (Cat. No. 4444556)

*Subject to terms and conditions. For complete details, go to www.thermofisher.com/taqmanguarantee.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 20X
Content And Storage 1 tube containing a 20X (S and M sizes) or 60X (L size) mix of pre-formulated assay (1 probe and 2 primers).

Store at -15 to -25°C.

Detection Method Primer-probe
Format Tube
GC-Rich PCR Performance High
PCR Method qPCR
Reaction Speed Fast
For Use With (Equipment) 7500 System, 7500 Fast System, 7900HT System, StepOne™, StepOnePlus™, ViiA™ 7 System, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 7 Flex, QuantStudio 6 Pro, QuantStudio 7 Pro, QuantStudio™ 12k Flex
Green Features Sustainable packaging
Product Line TaqMan
Product Type Gene Expression Assay
Quantity M (750 reactions), made to order
Shipping Condition Room Temperature
Fidelity (vs. Taq) 60 X
No. of Reactions 750 reactions
Show More Show Less
compliance-icons
Product Identifier
  • TAQMAN GENE EX ASSAYS MTO
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 2
  • Reproductive toxicity Category 1B
Hazard Statement
  • H351-Suspected of causing cancer.
  • H360-May damage fertility or the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : formamide

For Research Use Only. Not for use in diagnostic procedures.

Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt