Learn More
Applied Biosystems™ TaqMan™ GMO Maize 35S Detection Kit, with protocol
Kit contains reagents needed to reliably detect the presence of genetically modified organism (GMO)-specific DNA sequences in seed, grain, processed foods, and their ingredients.
£1085.00
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
12079800
|
Applied Biosystems™
4327690 |
1 Kit |
£1085.00
Each |
Estimated Shipment: 14-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Contains reagents needed to reliably detect the presence of genetically modified organism (GMO)-specific DNA sequences in seed, grain, processed foods, and their ingredients
- Universal assay detects and quantifies all approved genetically m