missing translation for 'onlineSavingsMsg'
Learn More

TaqMan™ TaqMan™ MicroRNA Reverse Transcription Kit

Product Code. 10268364 Shop All Applied Biosystems Products
Click to view available options
Sufficient For:
1000 Reactions
200 Reactions
Unit Size:
1000 reactions
200 reactions
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10268364

Brand: TaqMan™ 4366597

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Provides the necessary components for 200 reactions of optimal performance in TaqMan™ MicroRNA Assays.

A highly specific kit that quantitates only mature miRNAs, not precursors. A sensitive kit that conserves limited samples and requires only 1 to 10 nanograms of total RNA. It is fast, simple and scalable 2-step quantitative RT-PCR assay that provides high-quality results in less than three hours.

Components of this kit are used with the RT primer provided with the TaqMan™ MicroRNA Assay to convert miRNA to cDNA.

Order Info

Shipping Conditions: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Concentration 10X
Content And Storage Contains 10X RT buffer, dNTP mix w/dTTp (100 mM total), RNase inhibitor (20 U/μL), and MultiScribe™ RT enzyme (50 U/μL) for 1000 reactions. Store at -15 to -25°C.
Detection Method Primer-probe
GC-Rich PCR Performance High
PCR Method 2-step RT-qPCR
Reaction Speed Fast
Optimal Reaction Temperature 42°C
Product Line TaqMan
Product Type MicroRNA Reverse Transcription Kit
Quantity 1,000 reactions
Reverse Transcriptase MultiScribe™
Shipping Condition Dry Ice
Sufficient For 1000 Reactions
For Use With (Application) miRNA analysis
Fidelity (vs. Taq) 10 X
Final Product Type cDNA
No. of Reactions 1000 Reactions
Reaction Format Separate Components
Sample Type RNA
Size (Final Product) 200 bp or less
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.