missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ TaqMan™ Non-coding RNA Assay
£318.00 - £1042.00
Specifications
Content And Storage | 1 tube containing a 20X (S and M sizes) or 60X (L size) mix of pre-formulated assay (1 probe and 2 primers). Store at -15 to -25°C. |
---|---|
Green Features | Sustainable packaging |
Product Line | TaqMan™ |
Shipping Condition | Room Temperature |
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
11620129
|
TaqMan®
4426961 |
360 Reactions |
£318.00
Each |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11630129
|
TaqMan®
4426962 |
750 Reactions |
£510.00
Each |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11650129
|
TaqMan®
4426963 |
2900 Reactions |
£1042.00
Each |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
1 tube containing a 20X (S and M sizes) or 60X (L size) mix of pre-formulated assay (1 probe and 2 primers). Store at -15 to -25°C. |
|
Sustainable packaging | |
TaqMan™ | |
Room Temperature |