missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ TaqMan™ Protein Assays Buffer Kit
Shop All Applied Biosystems Products
Click to view available options
Quantity:
4000 Reactions
Unit Size:
4000 reactions
Description
TaqMan™ Protein Assays Buffer Kit contains the buffers required for preparing assay probes and use them in TaqMan Protein Assays experiments. The kit contains dilution buffers and a storage buffer, all of which are used in combination with the TaqMan™ Protein Assays Oligo Probe Kit.(4448549)
Specifications
Specifications
| Product Type | Kit, Assay Buffer |
| Detection Method | Primer-Probe Detection |
| For Use With (Equipment) | 7500 Fast System, 7500 System, 7900HT Fast System, 7900HT System, StepOnePlus™, Fast Mode |
| Content And Storage | TaqMan™ Protein Assays Buffer Kit contains the buffers required for preparing assay probes and is used in combination with the TaqMan™ Protein Assays Oligo Probe Kit.(4448549) |
| Hot Start | No |
| Label or Dye | FAM |
| Sample Type | Tissue, Cells |
| Shipping Condition | Dry Ice |
| Product Line | TaqMan |
| For Use With (Application) | Proximity Ligation Assay |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction