missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ TaqMan™ Protein Assays Oligo Probe Kit
TaqMan™ Protein Assays Oligo Probe Kit
Brand: Applied Biosystems™
Code : 50
Additional Details : Weight : 0.01000kg
Description
TaqMan™ Protein Assays Oligo Probe Kit contains the 5’ and 3’ Prox-Oligos required to prepare the assay probes for TaqMan™ Protein Assays experiments. The kit contains the oligonucleotide pair used in combination with the TaqMan™ Protein Assays Buffer Kit (4448571)Specifications
TaqMan™ Protein Assays Oligo Probe Kit contains the oligonucleotide pair used in combination with the TaqMan™ Protein Assays Buffer Kit (4448571) | |
qPCR | |
7500 Fast System, 7500 System, 7900HT Fast System, 7900HT System, StepOnePlus™, Fast Mode | |
TaqMan™ | |
Wet Ice | |
Not Target-Specific |
Primer-probe | |
Proximity Ligation Assay | |
FAM | |
Protein Assays Oligo Probe Kit | |
4000 Reactions | |
4000 Reactions |
For Research Use Only. Not for use in diagnostic procedures.