missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ TaqMan™ RNase P Control Reagents Kit
TaqMan™ RNase P Control Reagents Kit
Brand: Applied Biosystems™ 4316844
Code : 50
Additional Details : Weight : 0.02000kg
Description
The RNase P gene is a single-copy gene that encodes the RNA moiety for the RNase P enzyme. Applied Biosystems™ TaqMan™ RNase P Control Reagents were designed with limiting primer concentrations to be used as the endogenous reference in multiplex reactions. Multiplex PCR is the use of more than one probe and primer pair in the same tube. This kit is designed to perform a 5' nuclease assay with TaqMan™ Universal PCR Master Mix using the provided human genomic DNA as template.Comprehensive Guidelines Included
Applied Biosystems™ has developed a comprehensive set of guidelines to ensure success when using Applied Biosystems™ Real-Time PCR reagents and instrumentation. These guidelines are simple and easy to follow. Many traditional variables, such as magnesium chloride concentration and the thermal cycling protocol itself, have been standardized, greatly reducing assay development time.
Normalization Built-In
All Applied Biosystems™ Real-Time PCR reagent kits contain a passive internal reference to normalize non-PCR related fluorescence fluctuations. Normalizing with a passive internal reference minimizes well-to-well variability that can result from a variety of causes, such as pipetting errors and sample evaporation. The passive reference is also essential for accurate results when multiple probes (with different reporter dyes) are combined in a single tube.
Note: See user's manual or package insert for limited label license, and trademark information. For Research Use Only. Not for use in diagnostics procedures.
Specifications
Passive Internal Reference Standards | |
Human Genomic DNA | |
Low | |
Standard | |
VIC | |
Real Time PCR Reagents | |
Human | |
RNase P |
Control Primer-Probe Set(s) & Template(s) | |
Tube | |
Multiplex qPCR | |
TAMRA™ Quencher (3'), VIC™ (5') | |
TaqMan™ | |
Wet Ice | |
1000 Reactions | |
1000 Reactions |
For Research Use Only. Not for use in diagnostic procedures.