missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ TaqMan™ RNase P Detection Reagents Kit
TaqMan™ RNase P Detection Reagents Kit
Brand: Applied Biosystems™ 4316831
Code : 50
Additional Details : Weight : 0.00910kg
Description
The Applied Biosystems™ TaqMan™ RNase P Detection Reagents kit provides the components needed to detect and quantitate genomic copies of the human RNase P gene using the 5' nuclease assay. Human RNase P gene is a single-copy gene encoding the RNA moiety for the RNase P enzyme. This 5' nuclease assay employs TaqMan™ Universal PCR Master Mix or TaqMan™ PCR Core Reagents for polymerase chain reaction (PCR) using the provided human genomic DNA as template.Comprehensive Guidelines Included
Applied Biosystems™ has developed a comprehensive set of guidelines to ensure success when using Applied Biosystems™ Real-Time PCR reagents and instrumentation. These guidelines are simple and easy to follow. Many traditional variables, such as magnesium chloride concentration and the thermal cycling protocol itself, have been standardized, greatly reducing assay development time.
Normalization Built-In
All Applied Biosystems™ Real-Time PCR reagent kits contain a passive internal reference to normalize non-PCR related fluorescence fluctuations. Normalizing with a passive internal reference minimizes well-to-well variability that can result from a variety of causes, such as pipetting errors and sample evaporation. The passive reference is also essential for accurate results when multiple probes (with different reporter dyes) are combined in a single tube.
Note: See user's manual or package insert for limited label license, and trademark information. For Research Use Only. Not for use in diagnostics procedures.
Specifications
Passive Internal Reference Standards | |
Human Genomic DNA | |
Low | |
Standard | |
FAM | |
Real Time PCR Reagents | |
Human | |
RNase P |
Control Primer-Probe Set(s) & Template(s) | |
Tube | |
qPCR | |
TAMRA™ Quencher (3'), FAM (5') | |
TaqMan™ | |
Dry Ice | |
100 Reactions | |
100 Reactions |
For Research Use Only. Not for use in diagnostic procedures.