Learn More
Applied Biosystems™ TaqMan™ siRNA Assay
TaqMan™ siRNA Assay
Brand:  Applied Biosystems™ 4440879
Code : NEW
Additional Details : Weight : 0.03000kg
 View more versions of this product
Product Code. 15581074
  • £746.00 / Each
Estimated Shipment: 05-08-2024
to see stock.
Description
Description
TaqMan™ siRNA Assays, Made-to-Order
Specifications
Specifications
TaqMan™ siRNA Assay, 1 x 575μL RT primer/1 x 750μL TaqMan™ Assay 750 rxns, Medium, 20X RT Primer/20X TaqMan™ Assay Concentration, Store at –15 to –25°C
1 Each
750 Reactions
TaqMan™ siRNA Assay
20X RT primer/20X TaqMan™ assay
1 x 575μL RT primer/1 x 750μL TaqMan™ assay