Learn More
Applied Biosystems™ TaqPath™ 1-Step Multiplex Master Mix
TaqPath 1-Step Multiplex Master Mix is a fast, single-tube, 4X RT-qPCR mix that provides for sensitive, reproducible detection of up to four different RNA/DNA targets in a single multiplex reaction.
£169.00 - £3025.00
Specifications
Detection Method | Primer-probe |
---|---|
Format | Tube |
PCR Method | 1-step RT-qPCR, Multiplex qPCR |
Polymerase | Fast DNA Polymerase |
Reaction Speed | Fast or Standard |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15671832
|
Applied Biosystems™
A28526 |
5 x 1 mL |
£1628.00
Pack of 5 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15661832
|
Applied Biosystems™
A28525 |
0.5 mL |
£169.00
0.50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15681832
|
Applied Biosystems™
A28527 |
10 mL |
£3025.00
10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Particularly useful for RNA virus detection and high throughput gene expression workflows, the mix includes thermostable MMLV reverse transcriptase, dNTPs, UNG, MUSTANG PURPLE™ dye (passive reference), and thermostable Fast DNA polymerase, all in a single tube.
This single-tube, 4X format facilitates easy reaction set up - just add user-supplied assay and sample. With reproducible performance even in the presence of inhibitors, coupled with stringent quality control to minimize lot-to-lot variation, TaqPath One-Step Multiplex Master Mix, is a superior choice for your multiplex diagnostic testing or development needs.
TaqPath 1-Step Multiplex Master Mix is a fast, single-tube, 4X RT-qPCR mix that provides for sensitive, reproducible detection of up to four different RNA/DNA targets in a single multiplex reaction. Particularly useful for diagnostics and biopharmaceuticals with applications in viral pathogen detection and high throughput gene expression workflows, the mix includes thermostable MMLV reverse transcriptase, dNTPs, UNG, MUSTANG PURPLE dye (passive reference), and thermostable Fast DNA polymerase all in a single tube. TaqPath is manufactured under a strong ISO13485 quality management system and is designed to produce consistent product performance lot after lot.
This single-tube, 4X format facilitates easy reaction set up—just add user-supplied assay and sample (Figure 1). With reproducible performance even in the presence of inhibitors, coupled with stringent quality control to minimize lot-to-lot variation, TaqPath 1-Step Multiplex Master Mix is a superior choice for your multiplex diagnostic or bioprocess quality control testing and development needs.
Features of the TaqPath 1-Step Multiplex Master Mix include:
• High sensitivity—to detect low-copy targets with reproducible CT results
• Broad dynamic range—with both RNA and DNA targets
• Optimized for multiplexing—up to four targets simultaneously with MUSTANG PURPLE reference dye
• Tolerant of inhibitors—commonly found in clinical and biopharma samples
• Advanced manufacturing—production and process controls ensure lot-to-lot consistency
• Compliance Package—containing most frequently requested compliance documents, available below, commonly used to comply with internal quality systems and regulatory requirements in molecular diagnostic assay and drug development
High sensitivity
To enable reproducible detection of low-titer pathogen and transcripts, TaqPath 1-Step Multiplex Master Mix has been optimized as a higher-concentration 4X master mix that allows you to input more sample into each reaction, increasing sensitivity even in low-volume reactions. Figure 1 shows the consistency of results obtained when detecting 10 copy inputs of RNA target.
Wide dynamic range compatible with RNA and DNA
TaqPath 1-Step Multiplex Master Mix has been optimized to provide high specificity and dynamic range for use with both RNA and DNA targets. Since virology labs often test for both RNA and DNA viruses, TaqPath 1-Step Multiplex Master Mix is designed to use a single protocol to assay both types of nucleic acid. This input flexibility can help streamline the number of different workflows in your lab to improve efficiency. Figure 2 demonstrates the excellent PCR linearity across a 6-log RNA dilution series, while Figure 3 exhibits a 6-log linear dynamic range for both RNA and DNA targets.
Optimized for multiplexing
TaqPath 1-Step Multiplex Master Mix has been validated for multiplexing up to four targets simultaneously, allowing for additional targets and/or controls to be run simultaneously for efficiency or quality control purposes. Rather than including ROX dye as a passive reference, the TaqPath 1-step Multiplex Master Mix includes the longer emission wavelength MUSTANG PURPLE dye as a passive reference. This longer wavelength passive reference dye enables measurement of the JUN dye, or a similar emission wavelength dye, in the channel previously used to measure ROX dye. Figure 3 demonstrates TaqPath 1-Step Multiplex Master Mix performance in 4-plex reactions with both RNA and cDNA targets.
Inhibitor tolerant
Unlike other master mixes on the market, the unique proprietary formulation of TaqPath 1-Step RT-qPCR Master Mix allows robust performance even in the presence of PCR inhibitors typically found in biopharmaceutical, molecular diagnostics, and research applications. Optimized performance in the presence of such substances that can normally inhibit PCR, such as heparin, hematin, or EDTA, helps increase confidence when working with a variety of complex clinical or biopharma samples. TaqPath master mixes demonstrate increased tolerance of inhibitors compared to competitor mixes (Figure 4).
Manufacturing production and process controls
TaqPath 1-Step Multiplex Master Mix is manufactured under a strong quality management system that utilizes traceable quality raw materials and validated operating procedures. With established controls from purchasing through QC release, TaqPath product manufacturing is designed to produce consistent product performance lot after lot. The stringent QC is important for a variety of applications, making the master mix ideal for bioprocess quality control testing, biologics adventitious testing, molecular diagnostics, and research applications.
General Purpose Reagent manufactured under cGMP
TaqPath master mixes are general purpose reagents labeled “For Laboratory Use” and manufactured in an ISO 13485-certified and FDA-registered facility that adheres to cGMP principles.
Compliance Package
In an effort to efficiently support your compliance needs, frequently requested compliance documents are available here:
• Statement of Quality relating to the current good manufacturing practices (cGMP) to which TaqPath master mixes adhere
• Detailed lot-specific Certificate of Analysis (COA) (see below in Documents & Downloads section)
• Product Stability Statement
• Supplier Evaluation Support
-- Completed Self Assessment
-- ISO certificates by manufacturing site
• For EU Reach compliance-related inquiries, email 'LSGChemicalCompliance@thermofisher.com'
• BSE/TSE Statement
• Change Control and Notification Registration
Not finding the compliance document you’re looking for? Technical Support is here to help.
Specifications
Primer-probe | |
1-step RT-qPCR, Multiplex qPCR | |
Fast or Standard | |
TaqPath™ | |
Dry Ice | |
4X |
Tube | |
Fast DNA Polymerase | |
QuantStudio™ 6 Pro Real-Time PCR System or QuantStudio™ 7 Pro, QuantStudio™ 3 or 5, QuantStudio™ 6/QuantStudio™ 7 Flex, QuantStudio™ 12K Flex, QuantStudio™ Dx, ViiA™ 7, 7500 , 7500 Fast, 7500 Fast Dx | |
qPCR Master Mix | |
Gene Expression | |
DNA, RNA |