missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqPath™ qPCR Master Mix, CG
GREENER_CHOICE

Designed and all lots are functionally tested to help ensure lot-to-lot reproducibility for Ct consistency and dynamic range across a wide variety of assays.

Brand:  Applied Biosystems™ A16247

1124.75 GBP valid until 2025-03-28
Use promo code "24107" to get your promotional price.


 View more versions of this product

Product Code. 16268100

  • £2045.00 / Pack of 5

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Efficient and linear detection of up to 8 logs with gene expression or miRNA assays
  • Enables reliable detection of low-copy templates with reproducible CT results
  • Robust multiplexing performance with exogenous or endogenous targets
  • Engineered to retain consistent performance in preassembled reactions for up to 48 hours
  • Demonstrates increased tolerance to inhibitors compared to competitor mixes
  • Manufactured with stringent production and process controls to help ensure lot-to-lot consistency
  • Labeled “For Laboratory Use”
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store in refrigerator (2° to 8°C).
QuantStudio™ 12K Flex, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6, QuantStudio™ 7
ROX (Pre-mixed)
Real Time PCR (qPCR), Gene Expression, miRNA analysis
Primer-probe
2500 Reactions
5 x 5mL
DNA, cDNA
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

  • Multiple Manufacturers
    Fall in love with big savings
    Promo Code: 24107

    Expires2025-03-31

    New Offer
    Shop Now
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Applied Biosystems™ TaqPath™ qPCR Master Mix, CG > 5 x 5mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For laboratory use.