Learn More
Thermo Scientific™ TARGET DP 350μL insert vials
Use certified clean, low particle, low background vials for your high sensitivity LC/MS and GC/MS applications.
Brand:  Thermo Scientific™ CERT5000-73LV
Product Code. 15348046
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • 100 vials and 100 caps with pre-assembled septa
  • Reusable two-compartment trays protect vials and closures while keeping matching supplies together
  • Clear trays make it easy to keep track of available supplies without opening containers