missing translation for 'onlineSavingsMsg'
Learn More
Tartronic acid, 97%, Thermo Scientific Chemicals Product Code.: 11440197

Tartronic acid, 97%, Thermo Scientific Chemicals

Product Code. 11440197
0.1 g
Click to view available options
Quantity:
0.1 g
0.5 g
This item is not returnable. View return policy

Product Code. 11440197

Brand: Thermo Scientific Alfa Aesar B21640.77

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Tartronic acid is used as a reactant in the catalytic oxidation with air to form mesoxalic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tartronic acid is used as a reactant in the catalytic oxidation with air to form mesoxalic acid.

Solubility
Soluble in water and alcohol.

Notes
Store in a cool place. Incompatible with bases, oxidizing agents and reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80-69-3
Molecular Formula C3H4O5
Molecular Weight (g/mol) 120.06
MDL Number MFCD00004237
InChI Key ROBFUDYVXSDBQM-UHFFFAOYSA-N
Synonym tartronic acid, hydroxymalonic acid, 2-hydroxymalonic acid, propanedioic acid, hydroxy, tartronicacid, 2-tartronic acid, hydroxypropanedioic acid, 2-hydroxymalonate, malonic acid, hydroxy, hydroxymalonateShow MoreShow Less
PubChem CID 45
ChEBI CHEBI:16513
IUPAC Name 2-hydroxypropanedioic acid
SMILES C(C(=O)O)(C(=O)O)O

Specifications

Melting Point ∽151°C (decomposition)
Assay Percent Range 0.98
Beilstein 1209791
Merck Index 14,9073
Solubility Information Soluble in water and alcohol.
Formula Weight 120.06
Percent Purity 98%
Quantity 0.1 g
Chemical Name or Material Tartronic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10249867 Certificate TypeCertificate of Analysis Date12/04/2024 Catalog NumberB21640.77,B21640.02
Lot Number10245174 Certificate TypeCertificate of Analysis Date24/10/2023 Catalog NumberB21640.77,B21640.02
Lot Number10247363 Certificate TypeCertificate of Analysis Date18/07/2023 Catalog NumberB21640.77,B21640,B21640.02
Lot Number10242422 Certificate TypeCertificate of Analysis Date03/08/2022 Catalog NumberB21640.77,B21640.02
Lot Number10100297 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB21640.77,B21640.02

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tartronic acid, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.