Learn More
Tashiro's indicator solution, In ethanol, Honeywell Fluka™
In ethanol
Brand: Honeywell Fluka 36083-6X1L
Code : NEW
Additional Details : Weight : 4.74000kg Transport : UN number : 1170 Chem class : 3 Pack group : II
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Tashiro’s indicator solution | |
78°C | |
≤100.0000% | |
Glass Bottle | |
6 x 1 L |
In ethanol | |
≥90.0000% | |
Analytical Reagent | |
0.79 g/cm3 | |
12°C |