missing translation for 'onlineSavingsMsg'
Learn More

Tashiro's indicator solution, in methanol, Honeywell Fluka™

In methanol

Brand:  Honeywell Fluka 35325-1L

Code : NEW

Additional Details : Weight : 1.01000kg Transport : UN number : 1230 Chem class : 3 Pack group : II


Product Code. 15604940

  • £254.62 / 1 L
In Stock
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Specifications

Tashiro’s indicator solution
1 L
18°C (64.4°F)
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Tashiro

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : methanol, Methylthioninium chloride

SDS
Documents

Documents

Certificates
Special Offers

Special Offers