Learn More
Thermo Scientific Chemicals TBE running buffer (10X)
Commonly used in nucleic acid electrophoresis. TBE Buffer is used for polyacrylamide and agarose gel electrophoresis. It is ideally suited for DNA and RNA separation on longer run, higher voltage or amperage gels. TBE is used with non-denaturing or denaturing (7 M urea) gels.
Specifications
Chemical Name or Material | TBE running buffer (10X) |
---|---|
Physical Form | Liquid |
Packaging | Plastic bottle |
Solubility Information | It is soluble in water. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15455089
|
Thermo Scientific Chemicals
J62788.K2 |
1 L |
N/A
|
N/A | |||||
15465089
|
Thermo Scientific Chemicals
J62788.K3 |
2 L |
N/A
|
N/A | |||||
15475089
|
Thermo Scientific Chemicals
J62788.K7 |
4 L |
N/A
|
N/A | |||||
Description
TBE (Tris/Borate/EDTA) Buffer is commonly used in nucleic acid electrophoresis. TBE Buffer is used for polyacrylamide and agarose gel electrophoresis. It is ideally suited for DNA and RNA separation on longer run, higher voltage or amperage gels. TBE is used with non-denaturing or denaturing (7 M urea) gels. It is also routinely used for DNA automated sequencing gel.5 TBE can also be used for agarose gels but is not recommended for preparative gels for recovery of nucleic acids. Tris-Borate-EDTA buffer has been used for pulsed-field gel electrophoresis (PFGE).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
TBE running buffer (10X) | |
Liquid | |
Plastic bottle | |
It is soluble in water. |
Safety and Handling
P201-P202-P264b-P280i-P281-P302+P352-P305+P351+P338-P308+P313-P332+P313-P362-P501c
H315-H319-H360FD
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- TBE running buffer (10X)
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Reproductive toxicity Category 1B
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : Boric acid, Glycine,N,N-1,2-ethanediylbis[N-(carboxymethyl)-, disodium salt, dihydrate
RUO – Research Use Only