missing translation for 'onlineSavingsMsg'
Learn More

TBE Sample Buffer, 6X, Thermo Scientific™

£47.74 - £56.90

Specifications

Chemical Name or Material Sample Loading Buffer
Color Blue
Concentration 6X
Physical Form Liquid
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15493139
View Documents
Thermo Scientific Alfa Aesar
J60357.AC
25 mL
£47.74
25mL
Please to purchase this item. Need a web account? Register with us today!
15403149
View Documents
Thermo Scientific Alfa Aesar
J60357.AD
50 mL
£56.90
50mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Sample Loading Buffer
Blue
6X
Liquid
Videos
SDS
Documents

Documents

Product Certifications