Learn More
Tocris Bioscience™ TBK1 control PROTAC™ 4
Negative control for TBK1 PROTAC™ 3i (Cat. No. 7259)
Brand: Tocris Bioscience™ 7260/5
Additional Details : Weight : 0.00970kg
Description
TBK1 control PROTAC™ 4 is a negative control for TBK1 PROTAC™ 3i (Cat. No. 7259). Binds TBK1 with high affinity, but exhibits no significant degradation of TBK1. PROTAC™ is a registered trademark of Arvinas Operations, Inc., and is used under license.Specifications
(2S,4S)-1-((S)-18-(4-((5-Bromo-4-((3-(N-methylcyclobutanecarboxamido)propyl)amino)pyrimidin-2-yl)amino)phenoxy)-2-(tert-butyl)-4-oxo-6,10,15-trioxa-3-azaoctadecanoyl)-4-hydroxy-N-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide | |
5 mg | |
C53H74BrN9O9S |
2052306-31-5 | |
Active Degrader Negative Controls | |
98% |