missing translation for 'onlineSavingsMsg'
Learn More

TBS, Tris Buffered Saline, 1X Solution, pH 7.4, Molecular Biology, Fisher BioReagents

£44.50

Specifications

Chemical Name or Material Tris Buffered Saline
Name Note 1X Solution, pH 7.4
CAS 7647-14-5, 7447-40-7, 7647-01-0, 7732-18-5, 77-86-1
Boiling Point 100°C
Color Colorless
View More Specs

Products 1
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10467583
View Documents Promotion Details
Fisher BioReagents
BP2472-1
1 L Nalgene Poly Bottle
£44.50
1 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Tris Buffered Saline (TBS) is used in biochemical and molecular biology applications.
Specifications

Specifications

Tris Buffered Saline
7647-14-5, 7447-40-7, 7647-01-0, 7732-18-5, 77-86-1
Colorless
DNase free
TBS
1X Solution, pH 7.4
100°C
Liquid
Protease free
Molecular Biology
Videos
Safety and Handling

Safety and Handling

Recommended Storage : RT

SDS
Documents

Documents

Product Certifications