missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCEANC2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£263.49 - £392.00
Specifications
Antigen | TCEANC2 |
---|---|
Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 1-4 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500 |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18445342
|
Novus Biologicals
NBP2-13419-25ul |
25ul |
£263.49
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18096529
|
Bio-Techne
NBP2-13419 |
0.1 mL |
£392.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TCEANC2 Polyclonal specifically detects TCEANC2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
TCEANC2 | |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
127428 | |
This antibody was developed against a recombinant protein corresponding to the amino acids: TMLELPDQTKENLVEALQELKKKIPSREVLKSTRIGHTVNKMRKHSDSEVASLAREVYTEWKTFTEKHSNRPSIEV | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot 0.4 ug/ml, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 1-4 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
Polyclonal | |
Rabbit | |
Human | |
C1orf83, chromosome 1 open reading frame 83, FLJ32112, FLJ39169, RP4-758J24.3, transcription elongation factor A (SII) N-terminal and central domaincontaining 2 | |
TCEANC2 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only