Learn More
Thermo Scientific™ TCF-32 Continuous-Flow Rotors
Perform multi-liter processing applications that require high RCFs such as viral separations, particle isolations and vaccine production with the help of these rotors.
Brand:  Thermo Scientific™ 45820
 View more versions of this product
Product Code. 12141670
  • £78147.99 / Each
Explore more special offers
Description
Description
Perform multi-liter processing applications that require high RCFs such as viral separations, particle isolations and vaccine production with the help of Thermo Scientific™ TCF-32 Continuous-Flow Rotors.
Compatible with:
Thermo Scientific Sorvall WX Ultra Centrifuge Series
Ordering Information:
Core is required for operation. Must be purchased separately.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TCF-32 Continuous-Flow Rotor
940mL/430 mL
32,000 rpm
Thermo Scientific™ Sorvall™ WX ultraCentrifuges
Continuous Flow Rotor
102,000 x g
106/42
Autoclavable
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ TCF-32 Continuous-Flow Rotors > TCF-32 Rotor System