Learn More
MP Biomedicals™ TCH™ Defined Serum Replacement, 50X Concentrate
For long-term culturing of cells
Brand: MP Biomedicals™ 092020026
215.50 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.02000kg
Description
- TCH™ is a fortified, multipurpose serum replacement for long-term culturing of primarily human anchorage-dependent and suspension cells.
- A universal serum replacement that supports human and mammalian cells; completely free of virus, mycoplasma and other infectious agents; contains no growth factors, steroid hormones, extraneous proteins or phenol red; protein origin - human; Drug Master on file with the FDA.
- TCH™ Defined Serum Replacement is highly effective in the production of biologicals, as well as in culturing other mammalian cells.
- This serum replacement is effective in research situations such as studying the effects of growth factors or in the manufacturing of cell derived products.
Specifications
Clear, Pink | |
Solution | |
20 mL | |
Supplied as a 50X concentrate | |
6.8 to 7.2 |
Store at +4°C. | |
Serum Replacement | |
Sterile | |
Highly effective in the production of biologicals, as well as in culturing other mammalian cells. |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.