missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCS ERK 11e, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
Description
Potent and selective extracellular signal-related kinase 2 (ERK2) inhibitor (Ki values are <2, 395, 540 and 852 nM for ERK2, GSK-3, Aurora Kinase A and Cdk2 respectively). Potently blocks proliferation of HT29 cells (IC50 = 48 nM). Orally bioavailable.
Specifications
Specifications
CAS | 896720-20-0 |
Molecular Formula | C24H20Cl2FN5O2 |
Quantity | 10 mg |
Synonym | vx-11e, vtx-11e, vx 11e, 4-2-2-chloro-4-fluorophenyl amino-5-methylpyrimidin-4-yl-n-1s-1-3-chlorophenyl-2-hydroxyethyl-1h-pyrrole-2-carboxamide, vertex-11e, pyrimidylpyrrole, 11e, tcs erk 11e, vx-11e tcs erk 11e, 4-2-2-chloro-4-fluoroanilino-5-methylpyrimidin-4-yl-n-1s-1-3-chlorophenyl-2-hydroxyethyl-1h-pyrrole-2-carboxamide, 4-2-2-chloro-4-fluorophenyl amin-o-5-methyl-4-pyrimidinyl-n-1s-1-3-chlorophen-yl-2-hydroxyethyl-1h-pyrrole-2-carboxamide |
InChI Key | WUTVMXLIGHTZJC-OAQYLSRUSA-N |
SMILES | CC1=CN=C(N=C1C2=CNC(=C2)C(=O)NC(CO)C3=CC(=CC=C3)Cl)NC4=C(C=C(C=C4)F)Cl |
IUPAC Name | 4-[2-(2-chloro-4-fluoroanilino)-5-methylpyrimidin-4-yl]-N-[(1S)-1-(3-chlorophenyl)-2-hydroxyethyl]-1H-pyrrole-2-carboxamide |
Molecular Weight (g/mol) | 500.355 |
PubChem CID | 11634725 |
Formula Weight | 500.35 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'