Learn More
Thermo Scientific™ TDE Series -86°C Ultra-Low Temperature Chest Freezers, General Purpose
Part of the Standard Performance (STP) family, Thermo Scientific™ TDE Series ultra-low temperature chest freezers, powered by H-drive, feature three sizes, maximizing storage capacity from 3,645 up to 21,600 2mL vials. The TDE Series is a sustainable and reliable solution with ultimate sample security and operational savings for every lab.
£10086.34 - £14720.97
Specifications
Temperature Range -50°C to -86°C
Door Style Horizontal Solid
No. of Doors 1
Cabinet Material Cold-rolled Steel
Interior Stainless Steel
View More Specs
Products 3
Product Code Brand Holds Capacity (Metric) Certifications/Compliance Voltage Price Quantity & Availability  
Product Code Brand Holds Capacity (Metric) Certifications/Compliance Voltage Price Quantity & Availability  
17734579
View accessory products View Documents Promotion Details
Thermo Scientific™
TDEC06386FV
Standard 2 inch boxes (63) 85 L CE, ISO 13485, RoHS, REACH, EPA SNAP, UKCA, Prop 65 230 V
£10086.34
Each
17744579
View accessory products View Documents Promotion Details
Thermo Scientific™
TDEC25286FV
Standard 2 inch boxes (252) 360 L CE, ISO 13485, RoHS, REACH, EPA SNAP, UKCA, Prop 65 230 V
£12544.36
Each
17754579
View accessory products View Documents Promotion Details
Thermo Scientific™
TDEC39686FV
Standard 2 inch boxes (396) 566 L CE, ISO 13485, RoHS, REACH, EPA SNAP, UKCA, Prop 65 230 V
£14720.97
Each
Description
Show More
Specifications
Specifications
-50°C to -86°C
1
Stainless Steel
Capacitive Touch UI
One Port Hole
Single Hand Lift
Alarm, Recorder
Single ID
Smart View Compatible
H-drive
Horizontal Solid
Cold-rolled Steel
Painted Steel
Optional LN2 Backup, CO2 Backup, Chart Recorder
Manual
No
RS485, 4-20Ma, Dry Contacts, USB
Optional (Device Link Connect)
Single
R290/R170+R290 Mix
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Thermo Scientific™ TDE Series -86°C Ultra-Low Temperature Chest Freezers, General Purpose