missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hoefer™ TE Blotting Unit Replacement Parts and Accessories: Cassettes
For use with TE62 Standard Transfer Tank with Cooling Chamber
£115.97 - £175.00
Specifications
Includes | 1 cassette assembly, 2 Foam sponges 3mm thick and 1 Foam sponge 6mm thick |
---|
Product Code | Brand | For Use With (Equipment) | Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Type | Price | Quantity & Availability | ||||
12004807
|
Hoefer™
TE24 |
TE 22 Mini Tank Transfer Unit | Gel Cassette |
£115.97
Each |
Estimated Shipment: 14-08-2024
Log in to see stock. |
||||
10437884
|
Hoefer™
TE24 |
TE 22 Mini Tank Transfer Unit | Gel Cassette |
£174.00
Each |
Estimated Shipment: 07-08-2024
Log in to see stock. |
||||
12004757
|
Hoefer™
TE44H |
TE 42, TE 62 Transfer Tank | Hinged Cassette |
£127.72
Each |
Estimated Shipment: 14-08-2024
Log in to see stock. |
||||
10331095
|
Hoefer™
TE44H |
TE 42, TE 62 Transfer Tank | Hinged Cassette |
£175.00
Each |
Estimated Shipment: 16-09-2024
Log in to see stock. |
||||
Description
- Accessories and Replacement Parts for TE62
Specifications
1 cassette assembly, 2 Foam sponges 3mm thick and 1 Foam sponge 6mm thick |