Learn More
Hoefer™ TE Blotting Unit Replacement Parts and Accessories: Mylar Masks
For use with TE70X, TE77X, TE70XP and TE77XP Semi-Dry Transfer Units
Brand:  Hoefer™ TE74
 View more versions of this product
Product Code. 11524454
  • £54.07 / Pack of 4
Estimated Shipment: 22-11-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Comes in regular (16.5 x 18.5 cm) and large (23 x 27.5 cm) sizes
  • 4 pcs
Specifications
Specifications
TE 70 Semi Dry Transfer Unit
Mylar™ Mask