Learn More
Chemical Identifiers
CAS | 102-71-6 |
---|---|
Molecular Formula | C6H15NO3 |
Molecular Weight (g/mol) | 149.19 |
MDL Number | MFCD00002855 |
InChI Key | GSEJCLTVZPLZKY-UHFFFAOYSA-N |
Synonym | Triethanolamine buffer solution |
PubChem CID | 7618 |
ChEBI | CHEBI:28621 |
IUPAC Name | 2-[bis(2-hydroxyethyl)amino]ethanol |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15495959
|
Thermo Scientific Alfa Aesar
J63793.AK |
250 mL |
£89.40
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Triethanolamine (TEA), an emulsifier, surfactant and capping agent, may be used to study its toxicity profile and mechanisms. TEA may be used in various drug delivery constructs to improve drug solubility. TEA may be used to cap CdTe nanoparticles.Triethanolamine is found are liquid laundry detergents, dishwashing liquids, general cleaners, hand cleaners, polishes, metalworking fluids, paints, shaving cream and printing inks. Triethanolamine is also used as organic additive in the grinding of cement clinker. TEA has also been used in photographic (silver halide) processing. It has been promoted as a useful alkali by amateur photographers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsTriethanolamine (TEA), an emulsifier, surfactant and capping agent, may be used to study its toxicity profile and mechanisms. TEA may be used in various drug delivery constructs to improve drug solubility. TEA may be used to cap CdTe nanoparticles.Triethanolamine is found are liquid laundry detergents, dishwashing liquids, general cleaners, hand cleaners, polishes, metalworking fluids, paints, shaving cream and printing inks. Triethanolamine is also used as organic additive in the grinding of cement clinker. TEA has also been used in photographic (silver halide) processing. It has been promoted as a useful alkali by amateur photographers.
Solubility
Soluble in water.1 M, clear, colorless
Notes
Air Sensitive & Hygroscopic.Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from water, moisture, air, acids, metals, halocarbons.
Chemical Identifiers
102-71-6 | |
149.19 | |
GSEJCLTVZPLZKY-UHFFFAOYSA-N | |
7618 | |
2-[bis(2-hydroxyethyl)amino]ethanol |
C6H15NO3 | |
MFCD00002855 | |
Triethanolamine buffer solution | |
CHEBI:28621 |
Specifications
102-71-6 | |
0.2 M | |
Liquid | |
Odorless | |
Triethanolamine buffer solution | |
GSEJCLTVZPLZKY-UHFFFAOYSA-N | |
2-[bis(2-hydroxyethyl)amino]ethanol | |
7618 | |
TEA buffer soln. |
Colorless | |
8.0 | |
C6H15NO3 | |
MFCD00002855 | |
Soluble in water.1 M,clear,colorless | |
C(CO)N(CCO)CCO | |
149.19 | |
CHEBI:28621 |
Safety and Handling
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.