missing translation for 'onlineSavingsMsg'
Learn More

Tebbe reagent, 0.5M solution in toluene, Thermo Scientific Chemicals

£195.98 - £487.00

Specifications

Chemical Name or Material Tebbe reagent
Name Note 0.5M solution in toluene
CAS 67719-69-1, 108-88-3
Color Purple to Red
Density 0.9270g/mL
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10772263
View Documents Promotion Details
Thermo Scientific Acros
368970250
25 mL Glass Bottle
£195.98
25mL
Please to purchase this item. Need a web account? Register with us today!
10089001
View Documents
Thermo Scientific Acros
368971000
100 mL AcroSeal™ Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For C-C Bond Formation (Olefination), Lewis acid
Specifications

Specifications

Tebbe reagent
67719-69-1, 108-88-3
0.9270g/mL
C13H18AlClTi
08,83; 10,87; 11,52; 12,54; 13,71; 15,79; 16,70; 17,69
Bis(cyclopentadienyl-μ-chloro(dimethylaluminum-μ-methylenetitanium
284.62
0.5M solution in toluene
Purple to Red
4°C
MFCD00151575
0.927
Solubility in water: reacts.
Solution
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage. May cause damage to organs through prolonged or repeated exposure. May be fatal if swallowed and enters airways. May cause drowsiness or dizziness. Suspected of damaging fertility or the unborn child. Highly flammable liquid and vapor.

GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Use personal protective equipment as required. Avoid breathing dust/fume/gas/mist/vapors/spray. Do not breathe dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician. Do NOT induce vomiting. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Ground/Bond container and receiving equipment.

Danger

Product Identifier
  • Tebbe reagent, 0.5M solution in toluene

Signal Word
  • Danger

Hazard Category
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H361-Suspected of damaging fertility or the unborn child.
  • H373-May cause damage to organs.
  • H336-May cause drowsiness or dizziness.
  • H225-Highly flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P331-Do NOT induce vomiting.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only