Learn More
Cole-Parmer™ CB-200D-IB Electronic Ice Bucket
Keep your samples cold, clean, and dry with the CB-200D-IB Electronic Ice Bucket.
Marca: Cole-Parmer™ 24120-00
Code : 57
Dettagli aggiuntivi : Weight : 5.50800kg
Descrizione
- Waterless—no more wet melting ice or floating sample tubes
- Ceramic-coated beads maintain a 4°C temperature for the entire working day
- Holds approximately 60 x 1.5 mL microtubes
- 3 year warranty
- Previous part number FNOICE
Use the electronic ice bucket to keep your tubes clean and dry while incubating at sub-ambient temperatures for long periods of time. A removable bucket is filled with ceramic-coated beads that can maintain a constant temperature and support the sample tubes. To save time, the bucket of beads can be incubated in the refrigerator overnight and inserted into the unit before use. Reusable beads can be autoclaved at 273°F (134°C).
A temperature control ensures all contents in the ice bucket are kept at an even temperature, whether cold or warm. A dark plastic lid helps maintain a constant temperature and protects light sensitive samples. View the current temperature on a five-digit orange LED display in either °C or °F.
Please Note: Effective June 2022, the Techne Electronic Ice Bucket Model number FNOICE has now been rebranded Cole-Parmer under Model number CB-200D-IB. All the same great features and specifications, just with a new and improved look.
Specifica
Ice Bucket Sample Cooler | |
0.1°C | |
Orange LED | |
Microtubes | |
190 x 240 x 225 mm | |
225 mm | |
190 mm | |
240 mm | |
100/230 V, 50/60 Hz | |
100 to 230V, 50 to 60 Hz | |
4.5 kg |
0°C to 40°C | |
Ceramic-coated Beads | |
Chilling samples or reagents throughout the whole working day | |
Aluminum bucket, bag of ceramic-coated beads, dark plastic lid, and power cord. | |
8.9 in. | |
7.5 in. | |
9.4 in. | |
100/230 V | |
50/60 Hz | |
9.9 lb. | |
60 x 1.5 mL Microtubes |