Learn More
Cole-Parmer™ Essentials Cell Culturing Flask for BS-200 Series Cell Culture Stirring Systems
This Cell Culturing Flask is a complete culture vessel—flask, stir rod, and caps—to use with biological stirrers.
Brand: Cole-Parmer™ Essentials 21837-30
444.06 GBP valid until 2024-12-31
Use promo code "21939" to get your promotional price.
This item is not returnable.
View return policy
Description
- Completely sealed vessel is ideal for pathogenic materials
Culture vessels come calibrated and have two side ports and a central filling port. All culture vessel parts can be autoclaved at 121°C and 15 psi for 30 minutes. Silicone-encapsulated stir rods reduce the possibility of cells attaching to and growing on the surface.
Specifications
500 mL | |
8.1 in. | |
Autoclavable |
Borosilicate Glass | |
205 mm | |
100 mm |