Learn More
Medline™ Technico™ DIGIT-080 Sugar Pocket Refractometer
Brand:  Medline™ 8100.0800M
Product Code. 10644693
  • £396.00 / Each
Estimated Shipment: 04-12-2024
to see stock.
Explore more special offers
Add to basket
Specifications
Pocket Sugar Refractometer
DIGIT-080
1 Each
0 to 80 Brix
Zero calibration with distilled water at 20°