Learn More
Usbeck Laborgeraete™ Teclu Burner
Conduct experimentation at hotter temperatures with the Usbeck Laborgeraete™ Teclu Burner. It is available for use with natural gas and propane, with achievable temperatures up to 1600°C.
Brand:  Usbeck Laborgeraete™ 1210
 View more versions of this product
Product Code. 10671442
  • £36.90 / Each
Explore more special offers
Alert:
Burners according to DIN 30665 cannot be switched off completely. A small flame remains burning even if needle valve or stop cock are turned off. The gas flow shall be disconnected by means of the central stopcock in the laboratory.
Description
Description
  • Air regulation
  • Needle valve
  • Variant of Bunsen burner
Specifications
Specifications
Teclu Burner
17 mm (Head)
155 mm
1 Ea.
Natural Gas
14 mm
1520°C
260 g