missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TEK (TIE2) [Y1108F] Recombinant Human Protein
Human TEK (TIE2) Y1108F, C-terminal fragment, amino acids Q771-A1124 (as in NCBI/Protein entry NP_000450.2) with a Y1108F mutation, N-terminal GST-HIS6 fusion protein with a 3C cleavage site, expressed in Sf9 insect cells.
Brand: Invitrogen™ PV6231
This item is not returnable.
View return policy
Description
- Validated in LanthaScreen™ Kinase Binding Assay
- Protocols available for simple use and optimized performance
- Multiple pack sizes available for convenient use
Specifications
Enzyme and Protein Activity Assay, Industrial and Applied Science, Kinase Assay, Kinase Biology, Kinase Proteins and Expression Reagents, Pharma and Biopharma, Protein Assays and Analysis, Protein Biology, Target and Lead Identification and Validation | |
69.0kDa | |
Store at -68 to -85°C. Aliquot at first use to avoid multiple freeze-thaws. | |
aa 771-1124 | |
Recombinant |
50mM HEPES pH 7.5, 100mM NaCl, 5mM DTT, 15mM reduced glutathione, 20% glycerol | |
1mg | |
TEK, TIE-2 | |
Human | |
GST Tag, His Tag |