Learn More
Sampling Systems™ TeleScoop Water Sampler
The TeleScoop water sampler is an extendable sampler ideal for waters and liquids in hard-to-reach places. Its extendable rods reach up to 6 m in length, and the robust yet lightweight design means easy handling, even for the longest of rods.
£21.22 - £74.95
Specifications
Product Type Telescopic Rod
Material Aluminum
Autoclavable Not Autoclavable
Graduated No
Sterility Non-sterile
View More Specs
Products 7
Product Code Brand Length (Metric) Price Quantity & Availability  
Product Code Brand Length (Metric) Price Quantity & Availability  
17962112
View Documents
Sampling Systems™
5065A-1001
1000 mm
£25.54
Each
Estimated Shipment: 14-04-2025
to see stock.
Add to basket
17922032
View Documents
Sampling Systems™
5065A-1201
1200 mm
£21.22
Each
Estimated Shipment: 14-04-2025
to see stock.
Add to basket
17942032
View Documents
Sampling Systems™
5065A-2501
2500 mm
£24.56
Each
Estimated Shipment: 14-04-2025
to see stock.
Add to basket
17809366
View Documents
Sampling Systems™
5065A-2801
2800 mm
£40.53
Each
Estimated Shipment: 13-05-2025
to see stock.
Add to basket
17952032
View Documents
Sampling Systems™
5065A3001
3000 mm
£40.53
Each
Estimated Shipment: 14-04-2025
to see stock.
Add to basket
17962032
View Documents
Sampling Systems™
5065A-4501
4500 mm
£54.92
Each
Estimated Shipment: 14-04-2025
to see stock.
Add to basket
17972032
View Documents
Sampling Systems™
5065A-6001
6000 mm
£74.95
Each
Estimated Shipment: 14-04-2025
to see stock.
Add to basket
Description
Description
  • Dual locking system and multi angle adaptor elbow so that numerous attachments can be preset to angles for effective sampling
  • Attachments are quickly and easily changed without the need for using additional tools
  • Variety of compatible sampling accessories
  • Ideal for sampling from river banks, overhead gantries, etc.
Specifications
Specifications
Telescopic Rod
Not Autoclavable
Non-sterile
Aluminum
No
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Sampling Systems™ TeleScoop Water Sampler